Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Qualitative Filter Paper: Grade 6 Circles

Filters particles twice as fast as Qualitative Filter Paper Grade 5. Fisherbrand™ Qualitative Grade 6 Filters provide fine particle retention for boiler water analysis.

$15.56

Specifications

Format Circle
Grade Grade 6
Porosity Fine
Filtration Speed Herzberg 715 sec.
For Use With (Application) A standard grade filter for boiler water analysis.
View More Specs

Products 1
Catalog Number Diameter (Metric) Price Quantity & Availability  
Catalog Number Diameter (Metric) Price Quantity & Availability  
09-924-532
View Documents
70 mm
Pack of 100 for $15.56
Add to cart
 
Description

Description

Filters particles twice as fast as Qualitative Filter Paper Grade 5. Fisherbrand™ Qualitative Grade 6 Filters provide fine particle retention for boiler water analysis.

 

These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: 3μm
  • Porosity: Fine
  • Filtration speed: Herzberg, 710 sec.
  • Surface: smooth

Specifications

Specifications

Circle
Fine
A standard grade filter for boiler water analysis.
Cellulose
Grade 6
Herzberg 715 sec.
3μm
Medium to slow
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit