Learn More
MilliporeSigma™ Qualitative Filter Papers
Supplier:  MilliporeSigma™ FP10404700
 View more versions of this product
Catalog No. FP10404700
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
These 100% alpha-grade cotton cellulose fiber papers are ideal for clarifying and removing particles of 2.5μm and larger.
They're suitable for gravity and low suction filtration and can also be used in vacuum filtration systems. Available in 4.7 and 9.0cm diameters
1Filtration speed conditions. DIN 53137, time for 10mL to pass through filter; Herzberg, time for 100mL to pass through 10cm2 filter paper with 10cm constant head pressure of water.
2Wet-strengthened papers should not be used for Kjeldahl analyses.
Order Info
Come in boxes of 100.
Compliance
All papers meet ASTM E 832 for laboratory filter papers.
Specifications
Specifications
11μm
Clarifying liquids, separating precipitates, soil analysis and seed testing procedures, air pollution monitoring and gas detection.
Medium
4.7 cm
150 sec.
104
100 per box