Learn More
Cell Biolabs IncSupplier Diversity Partner QUANT CELL SENESCENCE KT 5X50  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner CBA2325
The Quantitative Cellular Senescence Assay measures Senescence Associated (SA) beta-galactosidase activity by flow cytometry or epifluorescence microscope. A fluorogenic substrate is added directly to cells in a 35 mm dish. 5x10 assays/kit.
Catalog No. 50-672-678