Learn More
Thermo Scientific™ Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing QC Set
Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Supplier: Thermo Scientific™ R4721000
Description
Thermo Scientific™ Quanti-Cult Plus Growth Promotion Testing Set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
Includes:
10 preserved organism vials and 10 rehydration vials
Contents:
Aspergillus brasiliensis ATCC™ 16404™
Bacillus subtilis ATCC™ 6633™
Candida albicans ATCC™ 10231™
Clostridium sporogenes ATCC™ 11437™
Pseudomonas aeruginosa ATCC™ 9027™.
Specifications
16404TM* | |
Quanti-Cult Plus | |
Organism Sets | |
100 Test/Set |
Growth Promotion Testing QC Set/5 | |
QC set | |
Quantitative QC | |
100 Tests/Set |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.