Learn More
Whatman™ Quantitative Grade Filter Papers, Ashless: PROMO OFFER
Suitable for gravity or low-suction (low-vacuum) filtration
Manufacturer:  Whatman™ 1441185PM
 View more versions of this product
Catalog No. 09-850-LPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
41
Circle
18.5cm
Gelatinous precipitates and coarse particles. Metallurgy: iron and aluminum hydroxides. Air pollution monitoring. Normal loading; limited wet strength
Filter Paper
Coarse
>20μmm