Whatman™ Quantitative Grade Filter Papers, Ashless: PROMO OFFER

Suitable for gravity or low-suction (low-vacuum) filtration

Manufacturer: Whatman™ 1442185PM

 View more versions of this product

Catalog No. 09-855-FPM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Diameter (Metric) 185mm
    Particle Retention >2.5μmm
    Format Circle
    Porosity Medium
    For Use With (Application) Highly retentive, for very fine precipitates. Normal loading; limited wet strength. Laboratory standard for critical gravimetric analysis
    Grade 42
    Material Filter Paper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit