Learn More
Whatman™ Quantitative Grade Filter Papers, Ashless: PROMO OFFER
Suitable for gravity or low-suction (low-vacuum) filtration
Manufacturer:  Whatman™ 1443150PM
 View more versions of this product
Catalog No. 09-856-MPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
43
Circle
150mm
Foodstuff and soil analyses, particle collection in air pollution monitoring for XRF analysis. Normal loading; limited wet strength
Filter Paper
Medium
>16μmm