Whatman™ Quantitative Grade Filter Papers, Ashless: PROMO OFFER

Suitable for gravity or low-suction (low-vacuum) filtration

Manufacturer: Whatman™ 1444150PM

 View more versions of this product

Catalog No. 09-860-EPM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Porosity Fine
    Diameter (Metric) 150mm
    Particle Retention >3μmm
    For Use With (Application) Retains fine particles, but somewhat thinner and faster than Grade 42. Normal loading; limited wet strength
    Grade 44
    Format Circle
    Material Filter Paper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit