Whatman™ Quantitative Grade Filter Papers, Ashless: PROMO OFFER

Suitable for gravity or low-suction (low-vacuum) filtration

Manufacturer: Whatman™ 1440-185PM

 View all options for this product

Catalog No. 09-845-GPM


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Grade 40
    Material Filter Paper
    Format Circle
    Porosity Medium
    Diameter (Metric) 18.5cm
    Particle Retention >8μmm
    For Use With (Application) Widely used, general-purpose gravimetric grade. Medium crystalline precipitates: calcium oxalate, well-digested barium. Normal loading; limited wet strength

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit