Learn More
Whatman™ Quantitative Grade Filter Papers, Hardened Ashless: PROMO OFFER
Maximum ash of 0.006%
Manufacturer:  Whatman™ 1541185PM
 View more versions of this product
Catalog No. 09-851-KPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
100/Pk.
Coarse
>20 to 25μmm
541
18.5cm
Tough, smooth surface. Coarse gelatinous precipitates. Useful for intermediate stages of gravimetric analysis