Learn More
Whatman™ Quantitative Grade Filter Papers, Hardened Ashless: PROMO OFFER
Maximum ash of 0.006%
Manufacturer:  Whatman™ 1540125PM
 View more versions of this product
Catalog No. 09-869-HPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
100/Pk.
Medium
>8μmm
540
12.5cm
Gravimetric analysis of metals in acid/alkali solutions. Collecting hydroxides after precipitation by strong alkalis. Normal loading; high wet strength.