Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR Master Mix v2 Non-distribution product as customer accommodation.

Applied Biosystems™ QuantStudio™ 3D Digital PCR Master Mix v2 is optimized for use with the QuantStudio™ 3D Digital PCR System.

$190.00 - $493.00

Specifications

For Use With (Equipment) QuantStudio™ 3 D
Product Line QuantStudio™
Shipping Condition Dry Ice
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
A26358 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
A26358
1.5 mL
Each for $190.00
Add to cart
 
A26359 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
A26359
5 mL
Each for $493.00
Add to cart
 
Description

Description

Specifically formulated with all reagents required to help ensure robust PCR amplification of single copy templates.

QuantStudio 3D Digital PCR Master Mix v2 requires use of the QuantStudio 3D™ Digital PCR 20K Chip Kit v2 and is fully-compatible with all QuantStudio™ 3D Digital PCR Instrument hardware and software. It is optimized for use with isolated DNA or cDNA, TaqMan™ assays, and the QuantStudio 3D Digital PCR Chip v2.

QuantStudio 3D Digital PCR MasterMix v2 is designed to deliver accurate, sensitive, and precise digital PCR data on QuantStudio 3D Digital PCR instruments when combined with associated reagents and QuantStudio™ 3D Analysis Suite™ software.

Specifications

Specifications

QuantStudio™ 3 D
QuantStudio™
Dry Ice
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit