Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR Reagent Kit

The QuantStudio™ 3D Digital PCR Reagent Kit is designed to confirm the performance of the QuantStudio™ 3D Digital PCR System either at the time of system installation or as part of future, user-defined system verification procedure using established Applied Biosystems™ consumables, reagents, control DNA, and reference assays.

Manufacturer:  Applied Biosystems™ A26360

Encompass

Catalog No. A26360

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.


Description

Description

The QuantStudio™ 3D Digital PCR Reagent Kit may also be used for training users on the QuantStudio™ 3D Digital PCR System workflow.

The QuantStudio 3D Digital PCR Reagent Kit contains CEPH control DNA, GLA and RNase P reference assays, QuantStudio™ 3D Digital PCR Master Mix v2, and nuclease free water. The kit requires use of QuantStudio 3D™ Digital PCR Chip Kit v2 and is fully-compatible with all QuantStudio 3D™ Digital PCR Instrument hardware and software.

Specifications

Specifications

• 1 mL CEPH Control DNA, 50 μg/mL, store at -20°C.
• GLA Reference Assay, store at -20°C.
• RNase P Reference Assay, store at -20°C.
• Nuclease-free water, store at -20°C.
• 1.5 mL QuantStudio 3D Digital PCR Master Mix v2, store at -20°C until first use and then store at 2–8°C.
QuantStudio™ 3D
Dry Ice
1 mL CEPH Control DNA, 50 μ g/mL, GLA Reference Assay, RNase P Reference Assay, Nuclease-free Water, 1.5 mL QuantStudio 3D Digital PCR Master Mix V2
QuantStudio™
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit