Learn More
MilliporeSigma™ Supelco™ Quartz Torch Body
$363.00 - $482.50
Specifications
For Use With (Application) Spectrometry
View More Specs
Products 4
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
11-100-8722
View Documents Promotion Details
MilliporeSigma™ Supelco™
GX3180830511EA
Agilent/HP 4500 and Agilent/HP 7500
Each for $482.50
Add to cart
11-100-8827
View Documents Promotion Details
MilliporeSigma™ Supelco™
GX3180801771EA
Agilent/Varian 700-ES Series Axial, Vista Axial
Each for $445.50
Add to cart
11-100-8739
View Documents Promotion Details
MilliporeSigma™ Supelco™
GX3180803531EA
Agilent/Varian 700-ES Series Axial, Vista Axial and Agilent/Varian Iberty Series II Axial
Each for $471.50
Add to cart
11-100-9039
View Documents Promotion Details
MilliporeSigma™ Supelco™
GX3180805411EA
Agilent/Varian 700-ES Series Radial, Vista Radial, Liberty 100/120/150/200/220
Each for $363.00
Add to cart
Description
Description
Specifications
Specifications
Spectrometry