Learn More
Invitrogen™ Qubit™ Protein Assay Kit Available on GSA/VA Contract for Federal Government customers only.
$106.00 - $321.00
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
Q33212 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
Q33212
N/A 500 assays
Each for $321.00
Add to cart
Q33211 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
Q33211
N/A 100 assays
Each for $106.00
Add to cart
Description
Description
The Qubit Protein Assay Kit is designed specifically for use with the Qubit Fluorometer. Using between 1 and 20 µl of your sample, this assay can quantitate samples ranging from 12.5 µg⁄ml to 5 mg⁄ml and exhibits low protein to protein variation. The assay is highly selective for proteins and is designed to be accurate in the presence of reducing reagents, but not in the presence of a large amount of detergent. Common contaminants, such as reducing reagents (DTT, β-mercaptoethanol), salts, free nucleotides, amino acids, solvents, or DNA, but not detergents, are well tolerated in the assay. Slight protocol modifications are required for other contaminants. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µL and 20 µL is acceptable), and read the concentration.
Which product to choose for fluorometric protein quantitation?
· For 1–20 samples: use this Qubit Protein Assay Kit with the Qubit Fluorometer
· For 20–2000 samples: use the Quant-iT Protein Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 µL thin-walled PCR tubes are required but not included.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.