Learn More
Invitrogen Qubit RNA HS Assay Kit
$320.00
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
Q32852
![]() ![]() |
Invitrogen
Q32852 |
N/A | 100 assays |
Each for $107.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Q32855
![]() ![]() |
Invitrogen
Q32855 |
N/A | 500 assays |
Each for $320.00
|
|
Description
The Qubit RNA HS Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples. The assay is highly selective for RNA and will not quantitate DNA, protein, or free nucleotides. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein, are well-tolerated in the assay. The assay kit is designed to be accurate for RNA sample concentrations between 250 pg/µL and 100 ng/µL. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µL and 20 µL is acceptable), and read the concentration using the Qubit Fluorometer.
Which product to choose for RNA HS (High Sensitivity) quantitation?
· For 120 samples: use this Qubit RNA HS Assay Kit with the Qubit Fluorometer
· For 202000 samples: use the Quant-iT RNA HS Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500uL thin-walled PCR tubes are required but not included.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.