Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quercetin dihydrate, 97%, Alfa Aesar™
Click to view available options
Quantity:
25g
100g
500g
Description
Applications
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.
Solubility
Soluble in DMSO (68 mg/ml at 25°C), Acetic Acid (1 mg/ml), Aqueous Alkali (1 mg/ml), water (<1 at 25°C), and ethanol (21 mg/ml at 25°C).
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.
Solubility
Soluble in DMSO (68 mg/ml at 25°C), Acetic Acid (1 mg/ml), Aqueous Alkali (1 mg/ml), water (<1 at 25°C), and ethanol (21 mg/ml at 25°C).
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 310°C to 315°C |
Quantity | 25g |
Assay Percent Range | 97% |
UN Number | UN2811 |
Beilstein | 317313 |
Merck Index | 14,8034 |
Solubility Information | Soluble in DMSO (68mg/ml at 25°C),Acetic Acid (1mg/ml),Aqueous Alkali (1mg/ml),water (<1 at 25°C),and ethanol (21mg/ml at 25°C). |
Formula Weight | 338.28 (302.25 Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | Quercetin Dihydrate |
Safety and Handling
GHS H Statement
H301-H341
P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 204-187-1
RTECSNumber : LK8950000
TSCA : Yes
Recommended Storage : Ambient temperatures