Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quercetin hydrate, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
849061-97-8
C15H10O7·xH2O
302.24
MFCD00149487
OKXFBEYCJRMINR-UHFFFAOYSA-N
quercetin hydrate,2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate,quercetinhydrate,quercetine sophoretin /,regid_for_cid_747934,regid_for_cid_16212154,3,3;,4;,5,7-pentahydroxyflavon,2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
16212154
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
C1=CC(=C(C=C1C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O)O)O.O
849061-97-8
C15H10O7·xH2O
302.24
MFCD00149487
OKXFBEYCJRMINR-UHFFFAOYSA-N
quercetin hydrate,2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate,quercetinhydrate,quercetine sophoretin /,regid_for_cid_747934,regid_for_cid_16212154,3,3;,4;,5,7-pentahydroxyflavon,2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
16212154
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
C1=CC(=C(C=C1C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O)O)O.O
Melting Point >300°C
Quantity 100g
Water 12% max. (K.F.)
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 94% min. (HPLC)
Beilstein 18, 242
Merck Index 14, 8034
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,acetone,pyridine,,glacial acetic acid
Formula Weight 302.24
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material Quercetin hydrate
Show More Show Less

GHS H Statement
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Danger

RTECSNumber : LK8950000