Learn More
BioVision IncSupplier Diversity Partner Quick Apoptotic DNA Ladder Detection Kit: Quick Apoptotic DNA Ladder Detection Kit: Fast, Easy & Sensitive Kit to Detect DNA Fragmentation in Apoptotic Cells within 90 minutes. Detection Method: Agarose gel electrophoresis. 50 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K12050
Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells. BioVision’s Quick Apoptotic DNA Ladder Detection Kit provides an easy and sensitive means for detecting DNA fragmentation in apoptotic cells. Unlike other commercially available kits that require 1-2 days to perform the procedure, the new detection method requires less than 90 minutes to prepare DNA, with neither extraction nor using columns. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9513