Learn More
Sklar InstrumentsSupplier Diversity Partner Quick Release Iris Hook, Sklar, 12 in., 3mm, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 908346
Quick Release Iris Hook, Sklar, 12 in., 3mm, Stainless Steel. The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue. It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length. This product is straight with a 3mm hook tip and an overall length of 12 inches.
Catalog No. 50-143-6875