Learn More
Thermo Scientific™ Quick Release Clamp
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Thermo Scientific™ Quick Release Clamp.
Manufacturer:  Thermo Scientific™ 715560
 View more versions of this product
Catalog No. 715560
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Thermo Scientific™ Quick Release Clamp.
Universal Cassette Clamps
  • For all types of commercially available cassettes
  • Silver model is for HM 325, HM 355S, and HM 340E models with serial numbers above 34000
Quick Release Clamp
  • For embedding rings
Zero Orientation Device
  • Positions specimen holder flat against the knife or blade (aligns X, Y axis)
Description & Specifications
Specifications
Quick Release Clamp
Quick-release clamp