Learn More
Thermo Scientific™ Quick Release Clamp
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Thermo Scientific™ Quick Release Clamp.
$1697.00
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
715020S
View Documents
Thermo Scientific™
715020S
Universal Cassette Clamp (silver)
Each for $1,697.00
Add to cart
Description
Description
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Thermo Scientific™ Quick Release Clamp.
Universal Cassette Clamps
  • For all types of commercially available cassettes
  • Silver model is for HM 325, HM 355S, and HM 340E models with serial numbers above 34000
Quick Release Clamp
  • For embedding rings
Zero Orientation Device
  • Positions specimen holder flat against the knife or blade (aligns X, Y axis)