Learn More
Agilent Technologies QUICKCHANGE II KL SDM KIT
Supplier: Agilent Technologies 200522
QuikChange II XL site-directed mutagenesis kit, Contains: PfuUltra High Fidelity DNA polymerase, 10X Reaction buffer, QuikSolution reagent, dNTP mix, Dpn I restriction enzyme, QuikChange XL control plasmid and control primers