Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Quickfit™ Polyacetal Joint Clips

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

$173.02

Specifications

For Use With (Equipment) Glassware with conical joints
Type Joint Clamp
Material Polyacetal
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Equipment) Color Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Color Price Quantity & Availability  
05-880F
View Documents
Cole-Parmer™
0684020
Glassware with conical joints Red Pack of 12 for $173.02
Add to cart
 
Description

Description

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
Specifications

Specifications

Glassware with conical joints
Joint Clamp
Polyacetal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit