Learn More
Quidel™ AmpliVue™ Bordetella Assay Kits Available on GSA/VA Contract for Federal Government customers only.
In vitro diagnostic test for the qualitative detection of Bordetella pertussis nucleic acids
Manufacturer:  Quidel Corporation M209
 View more versions of this product
Catalog No. 23-043-137
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Nasopharyngeal Swab
Reagent storage conditions: 2°C to 8°C Process Buffer and Reaction Tubes; 2°C to 30°C Amplicon Cartridge and Detection Cassette
Assay Kit