Learn More
Quidel AmpliVue™ HSV 1+2 Assay Kit
For qualitative detection and differentiation of Herpes simplex virus 1 and 2 (HSV1 and HSV2) nucleic acids isolated from cutaneous or mucocutaneous lesion of symptomatic patients
Manufacturer:  Quidel M210
Catalog No. 23-043-147
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Alert:
The AmpliVue™ HSV 1+2 Assay is not FDA-cleared for use with cerebrospinal fluid (CSF). The assay is not intended for prenatal screening.
Includes:
  • AmpliVue HSV 1+2 Assay: 16 x Detection cassettes, 16 x Amplicon cartridge, 16 x Dilution tubes, and 16 x Reaction tubes
  • HSV Starter kit: AV Heat Block 95°C, AV Heat Block 64°C, AV Rack, HSV 1+2 Assay Kit, and AV HSV 1+2/VZV controls
This item is not returnable. View return policy
Description
Description
The AmpliVue HSV 1+2 Assay detects and differentiates HSV-1 and HSV-2 DNA by a HDA reaction which simultaneously amplifies a HSV-1 specific sequence and a HSV-2 specific sequence in the presence of an internal control sequence. The amplicons are subsequently detected by a three line DNA test strip embedded in the cross-contamination-proof cassette.
Implement anywhere:
  • Can be performed with minimal bench space and without PCR equipment or virology isolation rooms
  • CLIA moderate complexity offers easy training
  • Sealed, disposable cartridge reduces risk of amplicon contamination and need for an isolation room
  • Scalable batch size
Fast and easy anytime:
  • Results in 1 hour with less than 5 minutes hands-on time
  • Intuitive workflow: can be run by any techician 24/7
  • Simple results interpretation
  • Lyophilized reagents extend shelf life, reducing cost and re-ordering
Operational Efficiency:
  • Definitive and differentiated results are obtained via lateral flow in disposable cassette that directly detects amplicon
  • Uses Helicase Dependent Amplification (HDA) technology for sensitivity and accuracy: DNA probes and primers for better specificity, mutiplex in single reaction tube
Specifications
Specifications
16 Tests/Kit
AmpliVue HSV 1+2 Assay Kit
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For in-vitro diagnostic use.