Learn More
Quidel Molecular HSV 1+2/VZV Control Set
For qualitative detection and differentiation of Herpes simplex virus 1 and 2 (HSV1 and HSV2) nucleic acids isolated from cutaneous or mucocutaneous lesion of symptomatic patients
Manufacturer:  Quidel M109
Catalog No. 23-043-148
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes:
  • HSV 1+2 Assay: 16 x Detection cassettes, 16 x Amplicon cartridge, 16 x Dilution tubes, and 16 x Reaction tubes
  • HSV Starter kit: AV Heat Block 95°C, AV Heat Block 64°C, AV Rack, HSV 1+2 Assay Kit, and AV HSV 1+2/VZV controls
This item is not returnable. View return policy
Description
Description
  • Molecular HSV 1+2/VZV controls
Specifications
Specifications
50 Reactions/Kit
Quidel Molicular HSV1+2/VZV Control Set
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For in-vitro diagnostic use.