Learn More
Quidel AmpliVue™ C. difficile Assay Kit Available on GSA/VA Contract for Federal Government customers only.
For qualitative detection and identification of toxigenic Clostridium difficile bacterial nucleic acids extracted from stool samples
Manufacturer:  Quidel M201
Catalog No. 23-043-140
Due to product restrictions, please to purchase or view availability for this product.
Includes: Detection Cassettes (1185001), Dilution Buffer (M5015): 1.8mL, Lysis Buffer (M5016): 1mL; Reaction tubes (M5017), Amplicon Cartridge (1215400) and Flocked Swabs (M5013)
This item is not returnable. View return policy
Description
Description
C. difficile is a major cause of antibiotic-associated diarrhea and colitis. The virulence of C. difficile is believed to be mediated by the production of two toxins (Toxin A and Toxin B). Both toxin genes genes (tcdA and tcdB, respectively) are located within a 19.6 Kb pathogenicity locus (PaLoc), along with 3 other gene products.
  • Easy-to-read disposable self-contained cassette-based molecular testing
  • Uniform pipetting volumes
  • Approximately 80 minutes of total assay time
  • Number of tests per kit: 16
  • Limit of detection: 4.2 CFU/assay
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
CE marked
Specifications
Specifications
Healthcare
Detection cassette, dilution buffer, lysis buffer, reaction tubes, amplicon cartridge, flocked swabs
C.difficile assay kit
16 Tests