Quidel™ QuickVue™ In-Line™ Strep A Test

Manufacturer: Moore Medical Corp 79227

 View more versions of this product

Catalog No. 19-898-620

  Check Availability
Add to Cart

    Specifications

    Product Type Control Swab
    Detectable Analytes Group A Streptococcal Antigen
    Includes 6 Positive and 6 Negative control swabs
    Quantity 12 Boxes
    Sample Type Throat swab
    Shelf Life 14 months
    Test Time 5 min.
    Clia Complexity Waived

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit