Moore Medical Quidel™ QuickVue™ One-Step hCG Urine Test

Manufacturer: Moore Medical Corp 67841

 View more versions of this product

Catalog No. 19-898-415

  Check Availability
Add to Cart

    Specifications

    Product Type hCG Urine control set
    Accuracy >99%
    Detectable Analytes hCG in Urine
    Includes Cassette, pipette, package insert and procedure card
    Quantity 2 Boxes
    Sample Size 3 drops
    Sample Type Urine
    Sensitivity 25mIU/mL
    Shelf Life 18 months
    Test Time 3 min.
    Clia Complexity Waived

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit