Learn More
Moore Medical Moore Medical Quidel™ QuickVue™ One-Step hCG Urine Test
Specifications
Accuracy >99%
Clia Complexity Waived
Detectable Analytes hCG in Urine
Includes Cassette, pipette, package insert and procedure card
Sample Size 3 drops
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
19-898-417
View Documents
Moore Medical
68029
25 Boxes hCG Urine test This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications
Specifications
>99%
hCG in Urine
3 drops
18 Months
Waived
Cassette, pipette, package insert and procedure card
25mIU/mL
3 min.