Learn More
Quidel Sofia™ hCG FIA
Kinetic Check™ feature provides operator the flexibility of upto 5 minutes before placing test cassette into Sofia for analysis
Manufacturer:  Quidel 20263
 View more versions of this product
Catalog No. 23-043-079
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Specific to monoclonal antibodies of beta subunit of hCG
  • Eliminates operator to operator variability and reduces transcription and billing errors
  • Automated results in three minutes
  • High accuracy and consistency
  • Prints results and/or sends to LIS for easy transfer of patient records
  • 20mIU/mL sensitivity in urine
  • Store at room temperature, 18 months shelf life from date of manufacture
Specifications
Specifications
hCG Starter Kit
9 x 50 Tests