Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia™ RSV FIA Immunofluorescence Kit

For rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens

Supplier:  Quidel 20265

 View more versions of this product

Catalog No. 23-043-082


Due to product restrictions, please to purchase or view availability for this product.


Includes: RSV FIA 25-test Kit Contents:
  • Individually Packaged Cassettes (25): Mouse monoclonal anti-RSV antibodies
  • Reagent Tubes (25): Lyophilized buffer with detergents and reducing agents
  • Reagent Solution (25): Vials with salt solution
  • Sterile Nasopharyngeal Swabs (25)
  • Large, Pink Fixed Volume Pipettes (25)
  • Small, Clear Fixed Volume Pipettes (25)
  • RSV Positive Control Swab (1): Swab is coated with non-infectious RSV antigen
  • Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Streptococcus C antigen
  • Package Insert (1)
  • Quick Reference Instructions (1)
  • QC Card (located on kit box)
  • Printer Paper (1)

Description

Description

 

  • Use with Sofia Analyzer only
  • Test nasopharyngeal swab and nasopharyngeal aspirate/wash specimens taken directly from symptomatic patients less than 19 years of age
  • When run in conjunction with Sofia Influenza A+B FIA, a single specimen may be used for both tests
  • Accurate results within 15 mins.
  • 25 tests per kit

 

Applications:   IVD

CLIA Complexity: This test is CLIA moderately complex for pediatric patients 7 to less than 19 years of age. It is CLIA waived for children less than 7 years of age.

Certifications

CE Marked; CLIA Complexity: Waived

Specifications

Specifications

Waived
Nasopharyngeal swab and nasopharyngeal aspirate/wash specimens
IVD
9 x 25 Tests
Respiratory syncytial virus nucleoprotein Antigens
Sofia™ RSV FIA Immunofluorescence Test Kit
Individually packaged cassettes 25, reagent tubes 25, reagent solution 25, sterile nasopharyngeal swabs 25, large, pink fixed volume pipettes 25, small, clear fixed volume pipettes 25, RSV positive control swab 1, negative control swab 1, package insert 1, quick reference instructions 1, QC card, printer Paper 1
>15 min.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit