Quidel™Sofia™ Strep A FIA CLIA-Moderate Available on GSA/VA Contract for Federal Government customers only.

For rapid qualitative detection of Group A Streptococcal antigens from throat swabs of symptomatic patients

Manufacturer: Quidel Corporation 20259

 View more versions of this product

Catalog No. 21-564-102

  Check Availability
Add to Cart

    Specifications

    Product Type Streptococcus A Starter Kit
    Detectable Analytes Group A Streptococcal Antigen
    For Use With (Equipment) Sofia analyzer
    Includes Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
    Quantity 9 Kits
    Sample Type Throat swab
    Storage Requirements Room temperature
    Test Time 5 min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit