Contec™ Quiltec™ I Wipes

Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.

Manufacturer: Contec™ QT199B

 View more versions of this product

Catalog No. 19-033-742C

  Check Availability
Add to Cart

    Specifications

    Product Type Quiltec™ I wipes
    Certifications/Compliance ISO Class 3 and 4
    Dimensions (L x W) 9 x 9 in. (23 x 23cm)
    For Use With (Equipment) Cleanroom
    Length (English) Exterior 9 in.
    Material Polyester
    Packaging Quantity 900/Cs.
    Ply 2
    Sterile No
    Thickness 2-Ply
    Width (English) Exterior 9 in.
    Width (English) 9 in.
    Width (Metric) 23cm
    For Use With (Application) Ideal for cleaning interior surfaces of process tools and other equipment, general purpose cleaning or product wipe down
    Length (English) 9 in.
    Length (Metric) 23cm

    • These sealed-edge wipes offer an extremely high sorptive capacity. Two layers of quilted polyester fabric are laser-cut and sealed together to create pockets where additional fluid can be trapped.
    • Two-ply quilted polyester construction creates durable wiper with exceptional absorbency—absorbs more than twice the capacity of a single-layer wiper
    • Extremely low in both particles and fibers—ideal for interior surface cleaning of process tools, wiping down products, or general-purpose cleaning

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit