Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldine, 95+%, Thermo Scientific Chemicals

Catalog Number p-190249
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
91-63-4
C10H9N
143.19
MFCD00006756
SMUQFGGVLNAIOZ-UHFFFAOYSA-N
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech
7060
2-methylquinoline
CC1=CC=C2C=CC=CC2=N1
91-63-4
C10H9N
143.19
MFCD00006756
SMUQFGGVLNAIOZ-UHFFFAOYSA-N
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech
7060
2-methylquinoline
CC1=CC=C2C=CC=CC2=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2.0°C
Color White to Yellow
Density 1.0580g/mL
Boiling Point 248.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6070 to 1.6120
Quantity 250 mL
Beilstein 20, 387
Specific Gravity 1.058
Merck Index 15, 8163
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in chloroform,ether and alcohol
Formula Weight 143.19
Percent Purity 95+%
Physical Form Crystalline Powder
Chemical Name or Material Quinaldine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doct
EINECSNumber 202-085-1
RTECSNumber UZ9625000
TSCA TSCA
Recommended Storage Product may discolor to brownish on storage

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit