Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldoyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
50342-01-3
C10H6ClNO
191.62
WFVMVMAUXYOQSW-UHFFFAOYSA-N
quinaldoyl chloride,2-quinolinecarbonyl chloride,quinaldyl chloride 97,quinaldyl chloride,chloroformylquinoline,2-quinolinecarbonyl chloride 9ci,quinoline-2-carboxylic acid chloride
2760547
quinoline-2-carbonyl chloride
C1=CC=C2C(=C1)C=CC(=N2)C(=O)Cl
50342-01-3
C10H6ClNO
191.62
WFVMVMAUXYOQSW-UHFFFAOYSA-N
quinaldoyl chloride,2-quinolinecarbonyl chloride,quinaldyl chloride 97,quinaldyl chloride,chloroformylquinoline,2-quinolinecarbonyl chloride 9ci,quinoline-2-carboxylic acid chloride
2760547
quinoline-2-carbonyl chloride
C1=CC=C2C(=C1)C=CC(=N2)C(=O)Cl
Assay Percent Range 94% min. (GC)
Formula Weight 191.62
Physical Form Crystalline Powder
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange to Brown
Melting Point 94°C to 97°C
Quantity 1g
Chemical Name or Material Quinaldoyl chloride

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger