Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Quinazoline, 98%

Click to view available options
Quantity:
1g
5g
253-82-7
C8H6N2
130.15
MFCD00006712
JWVCLYRUEFBMGU-UHFFFAOYSA-N
phenmiazine,1,3-benzodiazine,1,3-diazanaphthalene,5,6-benzopyrimidine,benzo a pyrimidine,chinazolin,unii-ub9qur18nl,ub9qur18nl,benzopyrimidine
9210
CHEBI:36621
quinazoline
C1=CC=C2C(=C1)C=NC=N2
253-82-7
C8H6N2
130.15
MFCD00006712
JWVCLYRUEFBMGU-UHFFFAOYSA-N
phenmiazine,1,3-benzodiazine,1,3-diazanaphthalene,5,6-benzopyrimidine,benzo a pyrimidine,chinazolin,unii-ub9qur18nl,ub9qur18nl,benzopyrimidine
9210
CHEBI:36621
quinazoline
C1=CC=C2C(=C1)C=NC=N2
Flash Point 106°C (224°F)
Quantity 5g
Melting Point 45°C to 48°C
Boiling Point 243°C
Merck Index 14,8053
Formula Weight 130.15
Percent Purity 98%
Chemical Name or Material Quinazoline