Alfa Aesar™ Quinazoline, 98%

Manufacturer: Alfa Aesar™ B2409403

Quantity 1g

 View more versions of this product

Catalog No. AAB2409403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 253-82-7
    Molecular Formula C8H6N2
    Formula Weight 130.15
    MDL Number MFCD00006712
    Merck Index 14,8053
    Chemical Name or Material Quinazoline

    Specifications

    Formula Weight 130.15
    Molecular Formula C8H6N2
    MDL Number MFCD00006712
    CAS 253-82-7
    Merck Index 14,8053
    Boiling Point 243°C
    Melting Point 45° to 48°C
    Quantity 1g
    Flash Point 106°C (224°F)
    Chemical Name or Material Quinazoline
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit