Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinidine Sulfate, Dihydrate, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
10 g
50 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6591-63-5
C40H54N4O10S
782.95
ZHNFLHYOFXQIOW-YKSBFIEBSA-N
bis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
O.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1
6591-63-5
C40H54N4O10S
782.95
ZHNFLHYOFXQIOW-YKSBFIEBSA-N
bis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
O.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1

QU105, 6591-63-5, (C20H24N2O2)2•H2SO4•2H2O

Spectrum™ Quinidine Sulfate, Dihydrate, USP is an antimalarial and antiarrhythmic drug used to treat or prevent many types of irregular heartbeats. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +275° to +288°
Water 4-5.5%
Ignition Residue 0.001
CAS Max % 1
Percent Purity 99 to 101%
Grade USP