Learn More
Accela Chembio IncSupplier Diversity Partner QUININE 100G  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY014630100G
QUININE, 100G, 130-95-0, MFCD00198096, 97+%, RETEST PERIOD: 540DAYS, LIGHT SENSITIVE/MOISTURE SENSITIVE/NITROGEN OR ARGON/+4 c
Catalog No. 50-648-485