Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine

Manufacturer:  Alfa Aesar™ A1045930

Quantity 250g

 View more versions of this product

Catalog No. AAA1045930


  Check Availability
Add to cart

Specifications

Description

Applications
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

130-95-0
324.42
91867
Quinine, Anhydrous, may cont. up to 5% dihydroquinine
C20H24N2O2
MFCD00198096
14,8061

Specifications

130-95-0
324.42
91867
≈177°C (dec.)
−166° (c=1 in ethanol)
99%, (total base)
C20H24N2O2
MFCD00198096
14,8061
250g
Quinine, Anhydrous, may cont. up to 5% dihydroquinine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VA6020000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit