Learn More
Sigma Organic Chemistry Quinoline-2 3-dicarboxy | 1G | 643-38-9 | MFCD00071756
Supplier: Sigma Organic Chemistry BBO0007421G

Quinoline-2 3-dicarboxy , 1G
About this Item:
CAS #: 643-38-9
MDL #: MFCD00071756
Molecular Weight: 217.18
UNSPSC Code: 12352200
Chemical Formula: C11H7NO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.