Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quinoline-3-boronic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
Specifications
Packaging | Glass Bottle |
Infrared Spectrum | Authentic |
Quantity | 25g |
Color | Brown to Orange or Pink |
Melting Point | 148.0°C to 155.0°C |
Assay Percent Range | 97% |
Formula Weight | 172.98 |
Physical Form | Powder |
Percent Purity | 97% |
Chemical Name or Material | Quinoline-3-boronic acid |
Safety and Handling
GHS Signal Word: Warning