Alfa Aesar™ Quinoline-6-boronic acid, 97%

Manufacturer: Alfa Aesar™ H52867MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52867MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 376581-24-7
    Molecular Formula C9H8BNO2
    Formula Weight 172.98
    MDL Number MFCD03093100
    Chemical Name or Material Quinoline-6-boronic acid

    Specifications

    Formula Weight 172.98
    Molecular Formula C9H8BNO2
    MDL Number MFCD03093100
    CAS 376581-24-7
    Health Hazard 1 Warning
    Melting Point 106° to 109°C
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Quinoline-6-boronic acid
    Assay 97%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit