Alfa Aesar™ Quinoline, 97%

Manufacturer: Alfa Aesar™ A1154536

Quantity 500g

 View more versions of this product

Catalog No. AAA1154536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91-22-5
    Molecular Formula C9H7N
    Formula Weight 129.16
    MDL Number MFCD00006736
    UN Number UN2656
    Beilstein 107477
    Merck Index 14,8068
    Chemical Name or Material Quinoline

    Specifications

    Formula Weight 129.16
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P361-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C9H7N
    Boiling Point 237°C
    CAS 91-22-5
    UN Number UN2656
    Health Hazard 1 Danger
    pH 7.3
    Health Hazard 2 GHS H Statement
    H311-H350-H341-H302-H315-H319
    Toxic in contact with skin.
    May cause cancer.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    MDL Number MFCD00006736
    Beilstein 107477
    Merck Index 14,8068
    Melting Point −15°C
    Quantity 500g
    Flash Point 101°C (214°F)
    TSCA TSCA
    Solubility Information More soluble in hot than cold water; Miscible with alcohol, ether
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE
    RTECS Number VA9275000
    Chemical Name or Material Quinoline
    Assay 97%

    Description

    Applications
    Quinoline is used as a chemical intermediate for making medicines and other organic compounds.  It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.

    Solubility
    More soluble in hot than cold water. Miscible with alcohol, ether

    Notes
    Dissolves sulfur, As{2}O{3}, phosphorus

    GHS H Statement
    H311-H350-H341-H302-H315-H319
    Toxic in contact with skin.
    May cause cancer.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P305+P351+P338-P361-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit