Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Quinoline, 98%

Manufacturer:  Alfa Aesar™ 43225A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43225A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

91-22-5
129.16
UN2656
14,8068
C9H7N
MFCD00006736
107477
Quinoline

Specifications

91-22-5
C9H7N
129.16
UN2656
14,8068
Colorless to pale yellow
−15°C
1kg
Light Sensitive & Hygroscopic
98%
Liquid
1.6260
MFCD00006736
107477
1.090g/mL
237°C
7.3
101°C (214°F)
Quinoline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H350-H341-H302-H315-H319
Toxic in contact with skin.
May cause cancer.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE

RTECSNumber : VA9275000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit