Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%

Click to view available options
Quantity:
5g
25g
64201-64-5
C9H9NO2
163.176
MFCD00149472
CUSWDTBBCIXCRB-UHFFFAOYSA-N
quinoline n-oxide hydrate,quinoline, 1-oxide, monohydrate,1-oxidoquinolin-1-ium hydrate,quinoline-n-oxide hydrate,quinoline-1-oxide hydrate,quinolinol, oxamethane,acmc-1be96,quinoline 1-oxide monohydrate,quinoline n-oxide monohydrate
152363
1-oxidoquinolin-1-ium;hydrate
C1=CC=C2C(=C1)C=CC=[N+]2[O-].O
64201-64-5
C9H9NO2
163.176
MFCD00149472
CUSWDTBBCIXCRB-UHFFFAOYSA-N
quinoline n-oxide hydrate,quinoline, 1-oxide, monohydrate,1-oxidoquinolin-1-ium hydrate,quinoline-n-oxide hydrate,quinoline-1-oxide hydrate,quinolinol, oxamethane,acmc-1be96,quinoline 1-oxide monohydrate,quinoline n-oxide monohydrate
152363
1-oxidoquinolin-1-ium;hydrate
C1=CC=C2C(=C1)C=CC=[N+]2[O-].O
Applications
Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Notes
Hygroscopic. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 25g
Flash Point >110°C (230°F)
Melting Point ~50°C
Beilstein 5989589
Formula Weight 145.16 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Quinoline N-oxide hydrate
Assay (dry wt.), water ≈20%

RTECSNumber : VC2340000