Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoxaline, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
91-19-0
C8H6N2
130.15
MFCD00006719
XSCHRSMBECNVNS-UHFFFAOYSA-N
benzoparadiazine,1,4-benzodiazine,phenopiazine,phenpiazine,quinazine,benzopyrazine,1,4-diazanaphthalene,benzo a pyrazine,1,4-naphthyridine,chinoxalin
7045
CHEBI:36616
quinoxaline
C1=CC=C2C(=C1)N=CC=N2
91-19-0
C8H6N2
130.15
MFCD00006719
XSCHRSMBECNVNS-UHFFFAOYSA-N
benzoparadiazine,1,4-benzodiazine,phenopiazine,phenpiazine,quinazine,benzopyrazine,1,4-diazanaphthalene,benzo a pyrazine,1,4-naphthyridine,chinoxalin
7045
CHEBI:36616
quinoxaline
C1=CC=C2C(=C1)N=CC=N2
Boiling Point 220°C to 223°C
Flash Point 98°C
Color Yellow to Brown
Melting Point 29°C to 34°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
Assay Percent Range 98+%
Beilstein 23, 176
Merck Index 15, 8195
Solubility Information Solubility in water: soluble. Other solubilities: miscible in alcohol,ether,benzene,acetone
Formula Weight 130.15
Physical Form Crystalline Low Melting Solid
Percent Purity ≥98.5%
Chemical Name or Material Quinoxaline
Show More Show Less