Alfa Aesar™ Quinuclidine, 97+%

Manufacturer: Alfa Aesar™ H5449806

Quantity 5g

 View more versions of this product

Catalog No. AAH5449806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-76-5
    Molecular Formula C7H13N
    Formula Weight 111.19
    MDL Number MFCD00006690
    UN Number UN2811
    Beilstein 103111
    Merck Index 14,8081
    Synonym 1-Azabicyclo[2.2.2]octane
    Chemical Name or Material Quinuclidine

    Specifications

    Formula Weight 111.19
    Molecular Formula C7H13N
    Quantity 5g
    TSCA TSCA
    MDL Number MFCD00006690
    UN Number UN2811
    Beilstein 103111
    Merck Index 14,8081
    Synonym 1-Azabicyclo[2.2.2]octane
    Health Hazard 1 Danger
    Melting Point 157° to 160°C
    Health Hazard 2 GHS H Statement
    H301-H310-H318-H315
    Toxic if swallowed.
    Fatal in contact with skin.
    Causes serious eye damage.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P361-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number CL5594625
    CAS 100-76-5
    Chemical Name or Material Quinuclidine
    Assay ≥97%

    Description

    Applications
    Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

    Solubility
    Soluble in alcohol, diethyl ether, water and organic solvents.

    Notes
    Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.

    GHS H Statement
    H301-H310-H318-H315
    Toxic if swallowed.
    Fatal in contact with skin.
    Causes serious eye damage.
    Causes skin irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P361-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit